Learn More
TARGETMOL CHEMICALS INC TAK-659 hydrochloride 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T420910MG
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TAK-659 hydrochloride (TAK-659) is a potent and selective inhibitor of spleen tyrosine kinase (SYK) with an IC50 value of 3.2 nM. It is selective against most other kinases, but potent toward both SYK and FLT3. Purity 99.63%
Catalog No. 50-287-8766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.