Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556581 Supplier Invitrogen™ Supplier No. PA556581
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YINFLVKVLG EQSLQQTGVA AQGNILGLFP QGPHLPGLED RTLLENYQVP SPGPSHHIP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 75%.

TAL2 is a helix-loop-helix protein. Translocations between this gene on chromosome 9 and the T-cell receptor beta-chain locus on chromosome 7 have been associated with activation of the T-cell acute lymphocytic leukemia 2 gene and T-cell acute lymphoblastic leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tal2
Gene Accession No. Q16559
Gene Alias bHLHa19; Class A basic helix-loop-helix protein 19; T cell acute lymphocytic leukemia 2; tail anomaly lethal 2; Tail2; TAL bHLH transcription factor 2; Tal2; TAL-2; T-cell acute lymphocytic leukemia 2; T-cell acute lymphocytic leukemia protein 2; T-cell acute lymphocytic leukemia protein 2 homolog; zgc:111884
Gene Symbols Tal2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TAL2. Recombinant protein control fragment (Product #RP-95725).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.