Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590041 Supplier Invitrogen™ Supplier No. PA590041
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAFAKLRKLI PTHPPDKKLS KNETLRLAMR YINFLVKVLG EQSLQQTGVA AQGNILGLFP QGPHLPGLED RTLLENYQVP SPGPSHHIP; Positive Samples: Rat spleen.

TAL2 is a helix-loop-helix protein. Translocations between this gene on chromosome 9 and the T-cell receptor beta-chain locus on chromosome 7 have been associated with activation of the T-cell acute lymphocytic leukemia 2 gene and T-cell acute lymphoblastic leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Tal2
Gene Alias bHLHa19; Class A basic helix-loop-helix protein 19; T cell acute lymphocytic leukemia 2; tail anomaly lethal 2; Tail2; TAL bHLH transcription factor 2; Tal2; TAL-2; T-cell acute lymphocytic leukemia 2; T-cell acute lymphocytic leukemia protein 2; T-cell acute lymphocytic leukemia protein 2 homolog; zgc:111884
Gene Symbols Tal2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 20 to the C-terminus of human TAL2 (NP_0054121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 685229
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.