Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Talabostat Mesylate
GREENER_CHOICE

Catalog No. 371910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-191-0 10 mg
37-195-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-191-0 Supplier Tocris Bioscience™ Supplier No. 3719/10
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inflammasomes Activators

Talabostat mesylate is a potent DPP IV inhibitor (IC50 values are <4, 4, 11, 310 and 560 nM at DPP4, DPP8, DPP9, DPP7 and FAP, respectively). Activates the inflammasome sensor protein NLRP1b and selectively triggers pyroptosis in monocytes and macrophages. Induces immune-mediated tumor regression in multiple mouse models of cancer. Orally bioavailable.

Specifications

Chemical Name or Material B-[(2R)-1-[(2S)-2-Amino-3-methyl-1-oxobutyl]-2-pyrrolidinyl]boronic acid methanesulfonate
CAS 150080-09-4
Synonym Val-boroPro, BXCL 701
Quantity 10 mg
Molecular Formula C9H19BN2O3.CH3SO3H
Formula Weight 310.18
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.