Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tali™ Cellular Analysis Slides

Catalog No. T10795
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Slides
500 Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10795 500 Slides
T10794 50 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10795 Supplier Invitrogen™ Supplier No. T10795
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer.

The Tali™ Cellular Analysis Slides are specifically designed for use with the Tali™ Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 μL of sample per chamber. Each box contains 50 slides.

Ready to Use for Your Convenience
Each Tali™ Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali™ Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Cellular Analysis Slide
Product Line Tali
For Use With (Equipment) Tali™ Image-Based Cytometer
Detection Method Fluorescence
Format Slide
Content And Storage Contains ten boxes of 50 slides. Store at room temperature.
Quantity 500 Slides
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.