Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Talin 1 Recombinant Rabbit Monoclonal Antibody (PSH09-95)
GREENER_CHOICE

Catalog No. MA562909
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA562909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA562909 Supplier Invitrogen™ Supplier No. MA562909
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Positive Control: HeLa cell lysate, HUVEC cell lysate, HepG2 cell lysate, MEF cell lysate, NIH/3T3 cell lysate, RAW264.7 cell lysate, C6 cell lysate.Tissue Specificity: Low tissue specificity.Subcellular Location: Cell projection, Cytoplasm, Cell surface, Cell junction.Sequence Similarities: 98% Mouse/Rat.Predicted band size: 270 kDa.

Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Talin 1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone PSH09-95
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TLN1
Gene Accession No. P26039, Q9Y490
Gene Alias ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1
Gene Symbols TLN1
Host Species Rabbit
Immunogen Recombinant protein within human Talin 1 aa 1,700-2,000/2,541.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21894, 313494, 7094
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.