Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Talin Monoclonal Antibody (8D4)
GREENER_CHOICE

Catalog No. PIMA547740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547740 Supplier Invitrogen™ Supplier No. MA547740
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 240 kDa* protein corresponding to the molecular mass of Talin on SDS-PAGE immunoblots of human A431, rat PC12, and rabbit fibroblast cells.

Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Talin
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8D4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene TLN2
Gene Accession No. P26039, P54939, Q71LX4, Q9Y490, Q9Y4G6
Gene Alias 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; KIAA1027; mKIAA0320; RGD1565416; talin 1; talin 2; talin-1; Talin-2; TLN; Tln1; Tln2
Gene Symbols TLN1, TLN2
Host Species Mouse
Immunogen Clone 8D4 was generated from purified chicken gizzard talin. The antibody recognizes an epitope within the rod domain of talin from many species, including human, rat, mouse, chicken, and frog.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21894, 313494, 315776, 395194, 415374, 70549, 7094, 83660
Target Species Human, Mouse, Rat, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.