Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talniflumate 98.0+%, TCI America™
SDP

Catalog No. T31011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31011G 1 g
T3101200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31011G Supplier TCI America Supplier No. T31011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66898-62-2
Molecular Formula C21H13F3N2O4
Molecular Weight (g/mol) 414.34
MDL Number MFCD00866135
InChI Key ANMLJLFWUCQGKZ-UHFFFAOYNA-N
Synonym talniflumate, somalgen, lomucin, talniflumato, talniflumate usan:inn, talniflumatum inn-latin, talniflumato inn-spanish, phthalidyl 2-3-trifluoromethylanilino nicotinate, 3-phthalidyl ester of 2-3-trifluoromethyl phenyl amino-3-pyridinecarboxylic acid, phthalidyl 2-alpha,alpha,alpha-trifluoro-m-toluidino nicotinate
PubChem CID 48229
IUPAC Name 3-oxo-1,3-dihydro-2-benzofuran-1-yl 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylate
SMILES FC(F)(F)C1=CC=CC(NC2=NC=CC=C2C(=O)OC2OC(=O)C3=CC=CC=C23)=C1

Specifications

Melting Point 168°C
Color Yellow
Quantity 1 g
Formula Weight 414.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Talniflumate
RTECSNumber QT2999200
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.