Learn More
Cayman Chemical -Talnolol 5mg
SDP
Supplier:  Cayman Chemical 161165 MG
Encompass_Preferred
A 5-LO inhibitor (IC50 15 nM) selective for 5-LO over 12-LO and 15-LO (IC50s 500 and 300 nM respectively) inhibits A23187-induced 5-LO product formation in isolated human PMNs monocytes and whole blood (IC50s 0.5 0.45 and 1.5 UM respectively) increases survival in a mouse model of PAF-induced lethal shock at 3.5 mg/kg reduces pleural levels of LTB4 in a rat model of carrageenan-induced pleurisy
Catalog No. 50-297-0027
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.