Learn More
Clontech Labs 3P TALON CELL THRU RESIN
SDP
Supplier:  Clontech Labs 3P 635509
Encompass
TALON His-Tag Purification Resin lets you prepare exceptionally pure his-tagged proteins from bacterial, mammalian, yeast, and baculovirus-infected cells, under native or denaturing conditions.
Catalog No. NC9770406
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.