Learn More
Cayman Chemical Tamoxifen, 10540-29-1, 500 mg
SDP
Supplier:  Cayman Chemical 13258500
Encompass_Preferred
Molecular Formula C26H29NO, Purity ≥95%, Formula Weight 371.5; A selective estrogen receptor (ER) modulator; an ER antagonist in breast tissue, effective in treating early breast cancer; an ER agonist in bone and blood vessels and a partial ER agonist in uterine tissues.
Catalog No. NC0472400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.