Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tamoxifen Free Base MP Biomedicals

Catalog No. ICN15673880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15673880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15673880 Supplier MP Biomedicals, Inc Supplier No. 215673880

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
Synonym tamoxifen, trans-tamoxifen, crisafeno, soltamox, tamoxifene, diemon, tamoxifeno, tamoxifenum, citofen, istubol
PubChem CID 2733526
ChEBI CHEBI:41774
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.