CAS RN 10540-29-1
Tamoxifen, 98%
CAS: 10540-29-1 Molecular Formula: C26H29NO Molecular Weight (g/mol): 371.52 MDL Number: MFCD00010454 InChI Key: NKANXQFJJICGDU-QPLCGJKRSA-N Synonym: tamoxifen,trans-tamoxifen,crisafeno,soltamox,tamoxifene,diemon,tamoxifeno,tamoxifenum,citofen,istubol PubChem CID: 2733526 ChEBI: CHEBI:41774 IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Tamoxifen, MP Biomedicals
CAS: 10540-29-1 Molecular Formula: C26H29NO Molecular Weight (g/mol): 371.52 MDL Number: MFCD00010454 InChI Key: NKANXQFJJICGDU-QPLCGJKRSA-N SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
R&D Systems™ Tamoxifen
A Tocris product, part of R&D Systems. Estrogen receptor partial agonist/antagonist
Tamoxifen Free Base MP Biomedicals
CAS: 10540-29-1 Molecular Formula: C26H29NO Molecular Weight (g/mol): 371.52 MDL Number: MFCD00010454 InChI Key: NKANXQFJJICGDU-QPLCGJKRSA-N Synonym: tamoxifen,trans-tamoxifen,crisafeno,soltamox,tamoxifene,diemon,tamoxifeno,tamoxifenum,citofen,istubol PubChem CID: 2733526 ChEBI: CHEBI:41774 IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Tamoxifen, 99.24%, MP Biomedicals™
CAS: 10540-29-1 Molecular Formula: C26H29NO Molecular Weight (g/mol): 371.52 MDL Number: MFCD00010454 InChI Key: NKANXQFJJICGDU-QPLCGJKRSA-N Synonym: tamoxifen,trans-tamoxifen,crisafeno,soltamox,tamoxifene,diemon,tamoxifeno,tamoxifenum,citofen,istubol PubChem CID: 2733526 ChEBI: CHEBI:41774 IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine SMILES: CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1