Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tamoxifen, MP Biomedicals

Catalog No. MP215673894 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215673894 5 g
MP215673880 100 mg
MP215673883 250 mg
MP215673891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215673894 Supplier MP Biomedicals, Inc Supplier No. 0215673894
Add to Cart
Add to Cart

Tamoxifen, White Powder

  • It has been used to facilitate the recombination of ect2flox allele in mouse organs.
  • It has also been used to study its effect on lipopolysaccharide (LPS)-induced microglial activation.
  • Protein kinase C inhibitor (IC50 = 50-200μM depending on assay conditions in MCF-7 cells or IC50 = 100μM in rat brain).
  • It is a prodrug that is metabolized to active metabolites 4-hydroxytamoxifen (4-OHT) and endoxifen by cytochrome P450 isoforms CYP2D6 and CYP3A4.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10540-29-1
Molecular Formula C26H29NO
Molecular Weight (g/mol) 371.52
MDL Number MFCD00010454
InChI Key NKANXQFJJICGDU-QPLCGJKRSA-N
SMILES CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C to 100°C
Color White
Loss on Drying 0.0021
Refractive Index KR5919600
Quantity 5 g
Synonym [Z]-1-[p-Dimethylaminoethoxyphenyl]-1,2-diphenyl-1-butene, (Z)-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine, ICI 47699, trans-Tamoxifen
Solubility Information Practically insoluble in water (< 0.01% at 20°C); soluble in methanol, ethanol, 2-propanol, propyleneglycol, chloroform (50mg/mL - clear, colorless to faint yellow solution) or DMSO
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Physical Form Powder
Assay 0.9935
storageNote1 Store at +4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.