Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Tamper Evident Attention Label
SDP

Catalog No. 18688003
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-688-003 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-003 Supplier Desco™ Supplier No. 06722
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Attention Label designed to identify if a package has been opened or tampered with.

  • ESD Susceptibility Symbol (RS-471) identifies the contents of the packaging as ESD susceptible
  • Tamper evident
  • Label will tear when removed to indicate that the package has been opened
  • Core: 3 in.

Specifications

Autoclavable Not Autoclavable
Adhesive Type Tamper-evident
Color Yellow
Material Paper
Sterility Non-sterile
Display Text Attention: Observe Precautions for Handling Electrostatic Sensitive Devices
For Use With (Application) Packages that are ESD susceptible
Latex Free Yes
Quantity 500/Roll
Dimensions (L x W) 2 x 2 in. (50.5 x 50.8 mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.