Learn More
Medchemexpress LLC Tamra alkyne, 5-isomer | 945928-17-6 | MFCD29912899 | 96.6% | 467.52 g/mol | C28H25N3O4 | 50 MG
Supplier: Medchemexpress LLC HYW42308050MG
TAMRA alkyne, 5-isomer is an alkyne-derivatized tetramethylrhodamine fluorescent probe for bioorthogonal labeling by click chemistry. It is designed for selective conjugation to azide-bearing biomolecules and enables enrichment, in-gel fluorescence detection, and identification of modified proteins.
- Alkyne functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
- Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN reagents.
- Enables enrichment and in-gel fluorescence detection of labeled proteins.
- High purity (~96.6%) with characterized molecular weight and formula.
- Supplied as the single 5-isomer to ensure consistent labeling performance.
- Available in multiple package sizes for research use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.