Learn More
Medchemexpress LLC Tamra azide, 5-isomer | 825651-66-9 | 98.6% | 512.56 | C28H28N6O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15185710MG
Encompass_Preferred
TAMRA azide, 5-isomer is a tetramethylrhodamine-based fluorescent azide reagent designed for conjugation via click chemistry. The azide functionality enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), allowing efficient labeling of biomolecules for fluorescence detection and imaging applications.
  • Fluorescent tetramethylrhodamine dye with orange emission.
  • Azide functional group enables CuAAC and SPAAC conjugation reactions.
  • Suitable for labeling proteins, peptides, nucleic acids, and small molecules.
  • High purity (98.6%) for reliable analytical and imaging results.
  • Solid material for preparation of concentrated stock solutions in organic solvents.
  • Molecular weight 512.56; molecular formula C28H28N6O4.
Catalog No. 50-002-85889
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.