Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tamsulosin hydrochloride
GREENER_CHOICE

Catalog No. 305010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-501-0 10 mg
30-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-501-0 Supplier Tocris Bioscience™ Supplier No. 3050/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective α1A antagonist

Tamsulosin hydrochloride is a selective α1A-adrenoceptor antagonist (pKi values are 9.97, 9.64 and 8.86 for α1A, α1B and α1D subtypes respectively). Decreases resting maximal urethral pressure with negligible effect on mean arterial blood pressure in vivo.

Chemical Identifiers

CAS 106463-17-6
Molecular Formula C20H29ClN2O5S

Specifications

Quantity 10 mg
Formula Weight 444.97
Purity >99%
Molecular Weight (g/mol) 444.971
Chemical Name or Material Tamsulosin hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.