Learn More
United Scientific™ Tandem Pulley
SDPSCI_ED
Catalog No. S15875 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Single Tandem Pulley
Catalog No. Product Type
S15875 Single Tandem Pulley
1 options
Catalog No. S15875 Supplier United Scientific™ Supplier No. PTAN01
Add to Cart
Add to Cart
Ideal for use in demonstrating simple machines.
  • Free-running triple tandem pulley with plastic sheaves.
  • Available in single, double, and triple sheaves.
  • Single is 50 mm in diameter.
  • Double is 40 mm and 50 mm in diameter.
  • Triple is 30 mm, 40 mm and 50 mm in diameter.
  • Designed with plated aluminum frame and hooks on both ends.
Specifications
Product Type Single Tandem Pulley
Material Aluminum
Warning: Cancer and reproductive harm