Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Tangent Galvanometer
SDPSCI_ED

Catalog No. S139478 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139478 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139478 Supplier United Scientific™ Supplier No. TNGV01
Add to Cart
Add to Cart
This item is not returnable. View return policy

Classic demonstration of how a current loop generates a magnetic field.

Classic demonstration of how a current loop generates a magnetic field.

  • Two coils of insulated wire, with 5 and 10 turns respectively, are wound around a 6 in. diameter plastic ring
  • The ring is mounted on a plastic base with 3 binding posts that allow connection to five, ten, or fifteen turns of wire
  • A magnetic compass (included) can be placed on the base to observe and measure deflection of the compass
  • An activity guide is included
  • Power source and connecting wires not included

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.