Learn More
Novus Biologicals™ Tangential Flow Filtration (TFF)-Large EV Isolation Adaptor
Shop All R&D Systems Products

Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Quantity | 2 Units |
| Sample Type | EV containing matrices (urine, cell culture supernatant, plasma, serum, plant extracts) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.