Learn More
Thermo Scientific™ Tango™ ADRA2A-bla U2OS DA Assay Kit
Description
The Tango™ ADRA2A-bla U2OS cells contain the human Adrenergic, Alpha-2A-, Receptor (ADRA2A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ ADRA2A-bla U2OS cells cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ ADRA2A-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
Specifications
Specifications
| Accession Number | NM_000681.2 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Division-Arrested Cells |
| Content And Storage | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: • Tango™ ADRA2A-bla U2OS DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • Tango™ ADRA2A-bla U2OS DA Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The Tango™ ADRA2A-bla U2OS DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Cytoplasmic Tail | Tail from AVPR2 GPCR |
| Detection Method | Fluorescence |
| Gene Symbol | ADRA2A |
| Population Status | Single Cell Clone Origin |
| Product Line | Tango |
| Quantity | 8 x 106 Cells |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.