Learn More
Medchemexpress LLC Taniborbactam hydrochloride | 2244235-49-0 | 100.0% | 462.18 g/mol | C19H30BCl2N3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY109124A5MG
Encompass_Preferred
Taniborbactam hydrochloride is a reversible, selective boronic acid-containing pan-spectrum β-lactamase inhibitor supplied as a research-grade solid for biochemical and antimicrobial studies. It is intended for preclinical evaluation of β-lactamase inhibition, resistance mechanisms, and combination therapy assays.
  • Reversible, selective boronic acid-based β-lactamase inhibitor.
  • Active against a broad range of β-lactamase enzymes.
  • Supplied as a solid in small pack sizes suitable for laboratory screening.
  • High purity appropriate for analytical and biological assays (≈99.97%).
  • Soluble in DMSO and compatible with common assay buffers.
  • Useful for studies of antibiotic potentiation and combination therapy.
Catalog No. 50-002-64535