Learn More
Tank Liner, Aegis™ 5-14, top and bottom drain
Description
Use Thermo Scientific Tank Liners for non-sterile fluid handling operations such as waste collection or powdered media or buffer hydration processes. Tank Liners are constructed with our Thermo Scientific Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility.
Features include:
• Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design for best fit and reliability
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components
Recommended for:
• Harvest—collection and storage of bulk non-sterile process liquids
• Collect—non-sterile process liquids for further processing
• Hydrate—powdered buffers, media, and media supplements
Specifications
Specifications
| Capacity (Metric) | 100 L |
| For Use With (Application) | Single-Use Bioprocessing |
| Quantity | 100 L |
| Tubing Style | TPE |