Learn More
Corning™ Tank Liners
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
$137.30 - $236.00
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Product Type | Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Product Type | Size | Price | Quantity & Availability | ||
MT9130020
|
Corning™
9130020 |
130 L | 45.72 cm | Non-Gusseted Tank Liner | 48 in. x 18 in. |
Each for $205.50
|
|
||
MT9130030
|
Corning™
9130030 |
200 L | 57.15 cm | Non-Gusseted Tank Liner | 22.5 in. x 36 in. (57.2 cm x 91.4 cm) |
Each for $236.00
|
|
||
MT9130035
|
Corning™
9130035 |
200 L | 55.88 cm | Gusseted Tank Liner | 22 in. x 36 in. (55.9 cm x 91.4 cm) |
Each for $137.30
|
![]() |
||
Description
- Made from low-density polyethylene (LDPE)
- Eliminate costs associated with cleaning and validation of tanks
- Decrease labor costs and increase turnaround time
- Eliminate risks associated with cross contamination
- Help extend life of reuseable tanks, resulting in reduced capital expenditures
- Gamma irradiated, individually packaged
- Choice of gusseted and non-gusseted available
- Available in many sizes
Specifications
LDPE | |
Cylindrical |