Learn More
Invitrogen™ TANK Polyclonal Antibody

Description
TANK Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | TANK |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Tank |
| Gene Accession No. | Q92844 |
| Gene Alias | C86182; E430026L09Rik; ITRAF; I-TRAF; LOW QUALITY PROTEIN: TRAF family member-associated NF-kappa-B activator; OTTHUMP00000200694; OTTHUMP00000200695; OTTHUMP00000200696; OTTHUMP00000200698; OTTHUMP00000200700; OTTHUMP00000200703; OTTHUMP00000200704; TANK; TANK-like protein; TRAF family member associated NFKB activator; TRAF family member-associated Nf-kappa B activator; TRAF family member-associated NF-kappa-B activator; TRAF family member-associated NFKB activator; TRAF interacting protein TANK; TRAF2; TRAF-interacting protein; zgc:153048 |
| Gene Symbols | Tank |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.