Learn More
R&D Systems™ Tankyrase 1 Chemiluminescent Activity Assay
Description
Includes
- Histone-coated 96-stripwell Microplate
- Anti-PAR Monoclonal Antibody
- HRP-conjugated Goat Anti-Mouse Antibody
- Tankyrase 1
- XAV939a
- Diluents & Buffers
- Unmodified NAD Substrate
- HRP Substrat
Tankyrase 1, also known as PARP5a, is a Poly (ADP-ribose) Polymerase (PARP) that associates with telomeres via its interaction with the telomeric-repeat binding factor TRF1. This assay utilizes a histone-coated 96-well microplate and an anti-poly (ADP-rib
- Amenable to high-throughput
- Highly sensitive: capable of measuring 0.1 mUnits of Tankyrase 1
- 3 hour assay time
- Provides full length Tankyrase 1 enzyme and unmodified NAD substrate
Specifications
Specifications
| Assay Sensitivity | 0.1mU |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | ELISA Kit |
| Sufficient For | 96 Tests |
| Target Species | Multi-species |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.