Learn More
Medchemexpress LLC Tanshinone I | 568-73-0 | MFCD00210563 | 98.8% | 276.29 g/mol | C18H12O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN013410MG
Encompass_Preferred
Tanshinone I is a natural diterpenoid research standard used as a phospholipase inhibitor and analytical reference for biochemical and preclinical studies. It inhibits human type IIA secreted phospholipase A2 (sPLA2) (IC50 = 11 μM) and rabbit cytosolic phospholipase A2 (cPLA2) (IC50 = 82 μM), and is supplied as a high-purity solid for laboratory use.
  • Natural diterpenoid research standard for biochemical assays.
  • Inhibits sPLA2 (human type IIA) and cPLA2 (rabbit) with reported IC50 values.
  • High purity suitable for analytical and research applications.
  • Supplied as a solid for flexible formulation and storage.
  • Commonly used in studies of inflammation and cardiovascular pathways.
Catalog No. 50-002-95977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.