Learn More
Apexbio Technology LLC DPN 1428-67-7 10mg
SDP
Supplier:  Apexbio Technology LLC B676310
Encompass_Preferred
DPN (1428-67-7) is a non-steroidal agonist targeting estrogen receptor (ER ) It is designed to selectively bind and activate ER thereby modulating ER -mediated signal transduction DPN exerts its biological activity primarily through selective agonistic activation of ER DPN demonstrates high binding affinity with an EC50 value of approximately 0 85 nM Based on these pharmacological properties DPN holds research potential in neuroprotection and the study of neuropathological conditions
Catalog No. 50-000-91728
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.