Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum ICP Standard, 10,000 ppm Ta in 2% HF, Ricca Chemical
SDP

Catalog No. PTA10KW50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PTA10KW-50 50 mL Natural Poly Bottle
PTA10KW-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PTA10KW-50 Supplier Ricca Chemical Company Supplier No. PTA10KW50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12022-02-5 , 7664-39-3 , 7732-18-5
Molecular Formula F7H8N2Ta
Molecular Weight (g/mol) 350.015
InChI Key RDNGDFQMYLJTJR-UHFFFAOYSA-I
PubChem CID 71311477
IUPAC Name diazanium;heptafluorotantalum(2-)
SMILES [NH4+].[NH4+].F[Ta-2](F)(F)(F)(F)(F)F

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Ta
CAS Min % 94.0, 1.89, 1.83
CAS Max % 98.0, 1.96, 1.9
Equivalence 1 mL = 10 mg Ta
Physical Form Liquid
Color Colorless
Type Standard
Quantity 50 mL
Chemical Name or Material Tantalum ICP Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.