Learn More
Sigma Organic Chemistry Tantalum(V) ethoxide | 10G | 6074-84-6 | MFCD00049785 | 99.98% trace metals basis
Supplier: Sigma Organic Chemistry 33911310G

Tantalum(V) ethoxide, 10G
About This Item:
Synonym(s): Pentaethoxytantalum; Pentaethyl tantalate; Tantalum ethylate; Tantalum pentaethoxide; Tantalum(V) ethoxide
Linear Formula: Ta(OC2H5)5
Molecular Weight: 406.25
CAS Number: 6074-84-6
MDL Number: MFCD00049785
UNSPSC Code: 12352300
Purity: 99.98% trace metals basis
Boiling Point: 155 C/0.01 mmHg (lit.)
Melting Point: 21 C (lit.)
Density: 1.566 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.487 (lit.)
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.