Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAOK1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557834
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7834 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7834 Supplier Bethyl Laboratories, Inc Supplier No. A300524AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

TAOK1 is a TAO type protein kinase. TAOK1 expression has been documented in rat brain. ESTs have been isolated from human normal adrenal, bone and breast and cancerous duodenum, cervix and germ cell.

Specifications

Antigen TAOK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TAOK1
Gene Accession No. Q7L7X3
Gene Alias FLJ14314, hKFC-B, hTAOK1, KFC-B, KIAA1361, kinase from chicken homolog B, MAP3K16, MARK Kinase, MARKK, microtubule affinity regulating kinase kinase, prostate-derived STE20-like kinase 2, prostate-derived sterile 20-like kinase 2, PSK2, PSK-2, serine/threonine protein kinase TAO1 homolog, STE20-like kinase PSK2, TAO1, TAOK1, thousand and one amino acid protein 1, thousand and one amino acid protein kinase 1
Gene Symbols TAOK1
Host Species Rabbit
Immunogen Between 400 and 450
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57551
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.