Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAP Monoclonal Antibody (53H8)
GREENER_CHOICE

Catalog No. 01674958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-958 Supplier Invitrogen™ Supplier No. MA15882
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-5882 detects TAP in human samples. MA1-5882 has been successfully used in Western blot, immunoprecipitation, immunofluorescence and ELISA procedures. MA1-5882 detects a band of approximately 68 kDa. The MA1-5882 immunogen is TAP. Detects a band of approximately 68 kDa. Store at 4°C for one month, or in small aliquots at -20°C for longer periods.

The constitutive transport element (CTE) of type D retroviruses serves as a signal of nuclear export for unspliced viral RNAs. TAP (also known as NXF1) mediates the export of CTE-containing simian type D retroviral RNAs through binding directly to the CTE. TAP is associated with a recognized mRNA export pathway and is a member of the multigene family of NXF proteins. NXF proteins belong to an evolutionarily conserved family of proteins, which are characterized by a leucine-rich-repeat domain (LRR) followed by a region known as the Nuclear Transport Factor 2 (NTF2)-like domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 53H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene NXF1
Gene Accession No. Q9UBU9
Gene Alias Mex 67 homolog; MEX67; Mex67h; modifier of vibrator 1; mRNA export factor TAP; Mvb1; Nuclear RNA export factor 1; nuclear RNA export factor 1 homolog; Nxf1; TAP; tip associated protein; tip associating protein; tip-associated protein; Tip-associating protein; Vdp
Gene Symbols NXF1
Host Species Mouse
Immunogen TAP from SP2/0 cells.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.