Learn More
Invitrogen™ TAP26 Polyclonal Antibody

Description
Immunogen sequence: NLKHLYLAEE ERHRKQARKV DHPLSEQVHQ PLLEEQCSID EPLFEDQCSF DQPQPEEQCI KTVNSFTIPK K Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 61%.
Specifications
Specifications
| Antigen | TAP26 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCDC59 |
| Gene Accession No. | Q9P031 |
| Gene Alias | 2300004H16Rik; BR22; Ccdc59; coiled-coil domain containing 59; coiled-coil domain-containing protein 59; D10Ertd718e; HSPC128; RGD1311912; TAP26; Thyroid transcription factor 1-associated protein 26; thyroid transcription factor 1-associated protein 26-like protein; TTF-1-associated protein 26; TTF-1-associated protein BR2; Unknown (protein for IMAGE:8047375) |
| Gene Symbols | CCDC59 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.