Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAPBPL Monoclonal Antibody (OTI1C9)
GREENER_CHOICE

Catalog No. PIMA526150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526150 Supplier Invitrogen™ Supplier No. MA526150
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAPBPL Monoclonal Antibody for Western Blot, IHC (P)

Tapasin, or TAPBP, is a member of the variable-constant Ig superfamily that links major histocompatibility complex (MHC) class I molecules to the transporter associated with antigen processing in the endoplasmic reticulum (ER). The TAPBP gene is located near the MHC complex on chromosome 6p21.3. TAPBPL is a member of the Ig superfamily that is localized on chromosome 12p13.3, a region somewhat paralogous to the MHC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAPBPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TAPBPL
Gene Accession No. Q9BX59
Gene Alias BC017613; TAP binding protein like; TAP binding protein related; TAP binding protein-like; tapasin-like; TAPASIN-R; tapasin-related protein; tapasin-related protein-like; TAP-binding protein-like; TAP-binding protein-related protein; Tapbpl; Tapbplr; TAPBPL-R; TAPBPR; TAPBP-R
Gene Symbols TAPBPL
Host Species Mouse
Immunogen Full length human recombinant protein of TAPBPL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55080
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.