Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPBPL Monoclonal Antibody (OTI2E11), TrueMAB™, OriGene
SDP

Catalog No. p-200012826
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9631 30 μL
50-167-6003 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9631 Supplier OriGene Supplier No. TA505539S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAPBPL Monoclonal antibody specifically detects TAPBPL in Human samples. It is validated for Western Blot

Specifications

Antigen TAPBPL
Applications Western Blot
Classification Monoclonal
Clone OTI2E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TAPBPL
Gene Accession No. Q9BX59
Gene Alias BC017613; TAP binding protein like; TAP binding protein related; TAP binding protein-like; tapasin-like; TAPASIN-R; tapasin-related protein; tapasin-related protein-like; TAP-binding protein-like; TAP-binding protein-related protein; Tapbpl; Tapbplr; TAPBPL-R; TAPBPR; TAPBP-R
Host Species Mouse
Immunogen Full length human recombinant protein of human TAPBPL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55080
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.