Learn More
Med Vet International Zoetis Revolution Brown for Dogs, 10.1-20 lbs, 6-pack
Supplier: Med Vet International RXREVBROW
Revolution (selamectin) Topical Parasiticide kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of American dog tick (Dermacentor variabilis) infestations in dogs. For use in dogs 6 weeks and older
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.