Learn More
Med Vet International Medical Tape 3M Transpore White White 3 inch X 10 Yard Plastic Nonsterile, 1534-3, 4/Box
Supplier:  Med Vet International 447641BX
Encompass_Preferred
The 3M Transpore white surgical tape provides secure adhesion to skin, devices and tubing It offers bi-directional tearing and is easy to use while wearing gloves This 10-yard long surgical tape is porous for breathability and comfort It provides a secure hold for up to 72 hours This tape resists moisture for placement onto damp skin The 3M Transpore medical tape is latex-free to minimize irritation Each box has 4 rolls of 3-inch tape
Catalog No. 50-282-8942
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.