Learn More
Med Vet International McKesson Blue Cast Tape, 4 Inch x 4 Yard, 115-4B, 10/Box
Supplier: Med Vet International 733699BX
Cast Tape McKesson 4 Inch X 12 Foot Fiberglass Blue Stretches to conform well to contours for a more comfortable better-fitting cast Porous weave allows the skin to breathe Water-activated resin lets you control working time according to application needs Lightweight for easy mobility
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.