Learn More
Electron Microscopy Sciences Tape Dispenser Model MD-350C, 15 Rolls, 3" Height

Supplier: Electron Microscopy Sciences 7705304

For use with 60-yard tape rolls. Made from durable polycarbonate for visibility and easy cleaning. Cutting edge is stainless steel and suction feet help hold in place. Spindles drop in for quickly changing out rolls.
Rolls: 15
Height: 3"
Width: 9 15⁄16"
Depth: 6 ¼"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.