Learn More
Med Vet International Steam Indicator Tape 3M Comply 3/4 inch X 60 Yard Steam, 1355-18mm, 28/Case
Supplier: Med Vet International 775749CS
For use as a process indicator to show that a pack has been exposed to the steam sterilization process Also serves to seal sterilization packs Can be used on disposable and reusable wraps Lead free ink means no hazardous waste issues Trusted, high quality adhesive Stretchable backing that minimizes tape "pop-off" during sterilization Can be written on or labeled with pre-printed labels
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.