Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Taped Tissue Cassette, 45° angle front
SDP

Catalog No. 22026811
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-026-811 Green
22-026-809 Aqua
22-026-810 Blue
22-026-813 Lilac
22-026-815 Pink
22-026-818 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-026-811 Supplier Azer Scientific Supplier No. ES6047G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tissue cassettes, taped for automation, hinged lid separate, 45° angle front, taped in stacks of 40, 50 stacks/case.

These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage. Molded from acetal, they keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.

This model differs by the special hinge that holds the base and cover together. The hinge design configuration allows for multiple opening and closing of the lid.

Features:

  • Made of acetal
  • Anterior writing area at a 45° angle
  • Easy-to-assemble disposable cover with special hinge
  • Large tab for convenient and easy opening
  • Covers packaged separately

Specifications

Closure Included Yes
Closure Type Lid
Color Green
Product Type Tissue Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.