Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TAPSO, for biochemistry

Catalog No. AC327810250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327810250 25 g Glass bottle
AC327811000 100 g Glass bottle
AC327815000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327810250 Supplier Thermo Scientific Chemicals Supplier No. 327810250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 68399-81-5 | C7H17NO7S | 259.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-81-5
Molecular Formula C7H17NO7S
Molecular Weight (g/mol) 259.273
MDL Number MFCD00038352
InChI Key RZQXOGQSPBYUKH-UHFFFAOYSA-N
Synonym 3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid
PubChem CID 109334
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO

Specifications

Absorbance (0.1 M aq. soln.)
Absorbance Conditions (0.1M aq. soln.)
Melting Point 220.0°C to 221.0°C
Color White to Yellow
pH 4.3 to 5.3 (1% aq. soln. at 25°C)
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNHCH2CH(OH)CH2SO3H
Packaging Glass bottle
Solubility Information Solubility in water: ca 129.6g/L (20°C).
Formula Weight 259.3
Percent Purity ≥99% (Acid)
Grade Biochemical
Chemical Name or Material TAPSO
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.