Learn More
MP Biomedicals™ Taq Core Kit 25
Catalog No. MP1EPTQK251 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 u
3 x 250 u
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1EPTQK251 250 u
MP1EPTQK253 3 x 250 u
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP1EPTQK251 Supplier MP Biomedicals™ Supplier No. 11EPTQK251
Add to Cart
Add to Cart
Taq DNA Polymerase + dNTPs in one kit
  • Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
  • MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
  • dNTPs at 25mM each
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 u
WARNING: Reproductive Harm - www.P65Warnings.ca.gov