Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq CORE Kit 25 Direct Loading

Catalog No. 11EPTQKL255 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 x 250U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11EPTQKL255 5 x 250U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11EPTQKL255 Supplier MP Biomedicals™ Supplier No. 11EPTQKL255

Includes

Taq DNA Polymerase 5U/μL (5 x 250U), 5X Direct Loading Buffer with Mg; 5X Direct Loading Buffer without Mg; dNTP mix (25mM each dNTP); 25mM MgCl2

Taq DNA Polymerase with dNTPs and direct loading buffers

The kits contain MP Biomedicals Taq DNA Polymerase (isolated from Thermus aquaticus), a thermo-stable recombinant polymerase, with a 5'-3' exonuclease activity, but no 3'- 5' exonuclease activity. These kits are also supplied with PCR grade dNTPs, as well as PCR Direct Loading Buffer 5X which contains incubation buffer for Taq DNA Polymerase, a densifying agent and a red-purple dye for direct loading onto agarose gel after PCR cycling. The red purple dye migrates at 400 bp on a 1.2% agarose gel.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage Taq DNA Polymerase 5U/μL (5 x 250U), 5X Direct Loading Buffer with Mg; 5X Direct Loading Buffer without Mg; dNTP mix (25mM each dNTP); 25mM MgCl2. Store at -20°C.
Gel Compatibility Agarose
Label or Dye Red-purple
Quantity 5 x 250U
For Use With (Application) Standard PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.