Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Taq DNA Ligase
SCI_ED

Catalog No. S000758
Change view
Click to view available options
Quantity:
10,000 U
2000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000758 2000 U
S000751 10,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000758 Supplier G-Biosciences Supplier No. 7862197
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taq DNA Ligase uses NAD+ as a cofactor to catalyze the formation of a phosphodiester bond in duplex DNA containing adjacent 5'-phosphoryl and 3'-hydroxyl termini.

It is active at high temperatures (50°C to 70°C) and used to detect a single-base substitution, allele-specific gene detection, mutagenesis during primer extension amplification.

Specifications

Packaging Vial
Enzyme DNA Ligase
Sensitive to Heat Inactivation No
Quantity 2000 U
Product Type Taq DNA Ligase
Research Category Traditional Cloning
Product Line Invitrogen
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.