Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq DNA Polymerase w/o Detergent

Catalog No. MP1EPTQX925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
10 x 250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP1EPTQX925 10 x 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1EPTQX925 Supplier MP Biomedicals™ Supplier No. 11EPTQX925

missing translation for 'validMainframeMsg'

Contains no detergent or BSA. MP Biomedicals Taq DNA Polymerase w/o Detergent is a specialized, thermostable enzyme. The incubation buffer (XD buffer) allows optimal results for researchers working with vegetal or beef material or using further dHPLC.

Contains a specialized, thermostable enzyme isolated from the thermophilic bacterium Thermus aquaticus. The recombinant form of this enzyme is expressed in E. coli.

  • Catalyzes oligonucleotide primer-driven, DNA-template-dependent incorporation of dNTPs into complimentary DNA strands
  • Displays both 5' to 3' polymerase and exonuclease activities
  • Used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences
  • Other application include RT-PCR, DNA sequencing and DNA labeling
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Freeze at −20°C
Form Liquid
Quantity 10 × 250 units
For Use With (Application) Standard PCR, RT-PCR, Sequencing, Nucleic Acid Labeling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.