Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq-&LOAD™ PCR Mastermix

Catalog No. 11EPTAL110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 100U
100U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11EPTAL110 10 x 100U
11EPTAL100 100U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11EPTAL110 Supplier MP Biomedicals™ Supplier No. 11EPTAL110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This mastermix allows direct loading after the PCR on the agarose gel without the need of loading dye.

Taq-&LOAD PCR Mastermix is a 5X concentrated solution containing MP Biomedicals recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer (includes a compound increasing the density of the sample and a red/purple dye). The included red/purple dye migrates at 400 bp on a 1.2% agarose gel.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage Recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer. Store at -20°C
Format Tube
Gel Compatibility Agarose
Label or Dye Red-purple
Quantity 10 x 100U
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.