Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Advanced miRNA Human A 96-well Plates, standard

Catalog No. A31811
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31811 4 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31811 Supplier Applied Biosystems™ Supplier No. A31811
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Applied Biosystems TaqMan Advanced miRNA Human A 96-Well Plates, Standard, provide the convenience of pre-spotted TaqMan Advanced miRNA assays in a 96-well standard plate format.

This product contains a set of 4 plates for profiling of up to 376 unique miRNAs, and includes endogenous and exogenous miRNA controls for normalization of data results.

Features of the TaqMan Advanced miRNA Human A 96-Well Plates include:

  • Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
  • Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
  • Updated content—assay content updated for mirBase v.21
  • Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
  • Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.

These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Detection Method Primer-probe
Format 96-well plate
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System, StepOnePlus™ System
Product Line TaqMan
Product Type Advanced miRNA Human 96-Well Plate
Quantity 4 plates
Shipping Condition Room Temperature
For Use With (Application) Microarray Analysis
Sample Type Liquid Samples (e.g. Serum), Tissue, Cells, FFPE & Fixed Samples, Blood
Volume 2 μL
Show More Show Less
What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.