Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TaqMan™ Array, Format 12

Catalog No. 4342247
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-422-47 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-422-47 Supplier LIFE TECHNOLOGIES Supplier No. 4342247
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Custom TaqMan® Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan® Gene Expression Assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C)
Format 12
Product Line TaqMan
Product Type Custom TaqMan Array Card
Quantity 1 each

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.